Learn More
Andwin Scientific SAFETY PIPETTER
SDP
Catalog No. NC9006300
Encompass_Preferred
Pipet Filler Handles pipettes from 2mL to 50mL. Comes with an adapter for capacities for more 20mL. Made of synthetic rubber. Color: red.
This item is not returnable. View return policy
Catalog No. NC9006300 Supplier Andwin Scientific Supplier No. 53502205
Pipet Filler Handles pipettes from 2mL to 50mL. Comes with an adapter for capacities for more 20mL. Made of synthetic rubber. Color: red.