Learn More
Caresfield Safety Push Button Butterfly Blood Collection Set
Description
- Push-button needle-retraction safety mechanism
- Large, ergonomic butterfly wings
- Double sheaths
- 200 per case
Specifications
Specifications
| Product Type | Safety Blood Collection Needle |
| Color | Orange |
| Material | Plastic, Stainless Steel |
| Needle Gauge | 25 G |
| Sterility | Sterile |
| Latex Free | Yes |