Learn More
Sklar Instruments Sklar Sterile Safety Scalpel, Sklar 11, BX10.
SDP
Supplier:  Sklar Instruments 063095
Encompass_Preferred
SklarSafe Sterile Safety Scalpel, Sklar, #11. The SklarSafe Sterile Safety Scalpel #11 features a protective, transparent sheath that can be opened, closed and locked with a single hand. The sturdy, in.sure-grip in. handle provides an audible in.click in. when the sheath is advanced to a locked position. This instrument is OSHA compliant and individually sealed in sturdy film and Tyvek pack. Safety scalpels are available in a box of 10.
Catalog No. 50-109-4403