Learn More
Accuform™ Safety Sign - CAUTION DO NOT UNPLUG, Adhesive Vinyl
Catalog No. 19119078 Shop All AccuformNMC Products
Catalog No. 19-119-078 Supplier Accuform™ Supplier No. MELC682VS
Add to Cart
Add to Cart
Accuform™ vaccination signs communicate health and safety policies, reinforce procedures, and identify locations
  • Printed matte surface has permanent acrylic adhesive back
  • Flexible vinyl can be applied to curved or uneven surfaces
  • Moisture resistant for various light-duty indoor and sheltered outdoor marking needs
  • Square corners and scored back liner for quick peel-off and application
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Specifications
    Material Vinyl
    Mounting Type Self-adhesive
    Shape Rectangular
    Languages English
    Displayed Text CAUTION DO NOT UNPLUG
    Color Red on Yellow