Learn More
Accuform™ Safety Sign: EMERGENCY EXIT ONLY
Catalog No. 17033241 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Polyester
Catalog No. Material
17-033-241 Polyester
1 options
Catalog No. 17-033-241 Supplier Accuform™ Supplier No. MEXT918VS
Add to Cart
Add to Cart
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage. They comply in accordance with OSHA 29 CFR 1910.145 and ANSI Z535 requirements.
  • Ideal for indoor and sheltered outdoor marking needs
  • Adhere to many flat or irregular surfaces
  • Flexible opaque poly with a printed matte surface
  • Permanent adhesive back with peel-off liner
Specifications
Certifications/Compliance OSHA 29 CFR 1910.145, ANSI Z535
Finish Non-glare
Material Polyester
Quantity 1 Sign
Adhesive Type Pressure-sensitive
Mounting Type Self-adhesive
Shape Rectangular
Languages English
Displayed Text EMERGENCY EXIT ONLY
Product Type Safety Sign
Color Red, White