Learn More
Accuform™ Safety Sign - ENTRANCE ONLY, Adhesive Vinyl
Catalog No. 19119179 Shop All AccuformNMC Products
Catalog No. 19-119-179 Supplier Accuform™ Supplier No. MADM434VS
Add to Cart
Add to Cart
Accuform™ signs identify/indicate potential, type and degree of hazards that may lead to accidental injury/property damage
  • Printed matte surface has permanent acrylic adhesive back
  • Flexible vinyl can be applied to curved or uneven surfaces
  • Moisture resistant for various light-duty indoor and sheltered outdoor marking needs
  • Square corners and scored back liner for quick peel-off and application
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Specifications
    Material Vinyl
    Mounting Type Self-adhesive
    Shape Rectangular
    Displayed Text ENTRANCE ONLY
    Color Red on White