Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Safety Sign - EXIT, Adhesive Vinyl

Catalog No. 19169800 Shop All AccuformNMC Products
Catalog No. 19-169-800 Supplier Accuform™ Supplier No. MADC531VS
Add to Cart
Add to Cart

Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage

  • Printed matte surface has permanent acrylic adhesive back
  • Flexible vinyl can be applied to curved or uneven surfaces
  • Moisture resistant for a various light-duty indoor and sheltered outdoor marking needs
  • Square corners and scored back liner for quick peel-off and application
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements

Specifications

Material Vinyl
Mounting Type Self-adhesive
For Use With (Application) Identifying potential hazards
Shape Rectangular
Displayed Text EXIT
Color Red on White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.