Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Safety Sign - EXIT, Aluminum

Catalog No. 19169805 Shop All AccuformNMC Products
Catalog No. 19-169-805 Supplier Accuform™ Supplier No. MEXT906VA
Add to Cart
Add to Cart

Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage

  • Thick, rigid aluminum has the strength of metal without rust
  • Printed surface has clear UV protective overcoat
  • Withstand splashes, wash-downs and outdoor conditions
  • Abrasion and chemical resistance
  • Rounded corners and 0.1875 in. mounting holes for installation
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements

Specifications

For Use With (Application) Identifying potential hazards
Shape Rectangular
Displayed Text EXIT
Product Type Safety Sign
Color Red on White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.