Learn More
Accuform™ Safety Sign - GET VACCINATED HERE, Plastic
Catalog No. 19119085 Shop All AccuformNMC Products
Catalog No. 19-119-085 Supplier Accuform™ Supplier No. MBDX515VP
Add to Cart
Add to Cart
Accuform™ vaccination signs communicate health and safety policies, reinforce procedures, and identify locations
  • Durable plastic has slight flex ideal for indoor and sheltered outdoor environments
  • Printed surface has clear UV protective overcoat
  • 50% recycled content with UV inhibitors and good impact resistance
  • Rounded corners with 0.1875 in. mounting holes for installation
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Specifications
    Material Polyethylene
    Mounting Type Corner Holes
    Shape Rectangular
    Languages English
    Displayed Text GET VACCINATED HERE
    Color Blue on White