Learn More
Accuform™ Safety Sign - NOTICE AVOID CONTAMINATION WASH YOUR HANDS, Aluminum
Catalog No. 19119172 Shop All AccuformNMC Products
Catalog No. 19-119-172 Supplier Accuform™ Supplier No. MCHL810VA
Add to Cart
Add to Cart
Accuform™ signs alert and remind others to follow health and safety policies to prevent spread of germs and viruses
  • Thick, rigid aluminum has strength of metal without rust
  • Printed surface has clear UV protective overcoat
  • Withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance
  • Rounded corners and 0.1875 in. mounting holes for installation
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Specifications
    Mounting Type Corner Holes
    Shape Rectangular
    Languages English
    Color Blue on White