Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Safety Sign - NOTICE Avoid Contamination WASH YOUR HANDS, Plastic

Catalog No. 19169798 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Polyethylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-169-798 Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-169-798 Supplier Accuform™ Supplier No. MCHL810VP
Add to Cart
Add to Cart

Signs alert and remind people to follow health and safety policies to prevent the spread of germs and viruses

  • Durable plastic has slight flex, ideal for indoor and sheltered outdoor environments
  • Printed surface has clear UV protective overcoat
  • 50% recycled content with UV inhibitors and good impact resistance
  • Rounded corners with 0.1875 in. mounting holes for installation
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements

Specifications

Certifications/Compliance Complies with OSHA 29 CFR 1910.145 and ANSI Z535
Material Polyethylene
Mounting Type Corner Holes
For Use With (Application) Reminding people to follow health and safety policies indoors/outdoors
Shape Rectangular
Languages English
Displayed Text NOTICE Avoid Contamination WASH YOUR HANDS
Product Type Safety Sign
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.