Learn More
Accuform™ Safety Sign - NOTICE FOR THE HEALTH AND SAFETY OF ONE ANOTHER FACE MASK REQUIRED, Adhesive Vinyl
Catalog No. 19119159 Shop All AccuformNMC Products
Catalog No. 19-119-159 Supplier Accuform™ Supplier No. MPPA836VS
Add to Cart
Add to Cart
Accuform™ signs alert and remind others to follow health and safety policies to prevent spread of germs and viruses
  • Printed matte surface has permanent acrylic adhesive back
  • Flexible vinyl can be applied to curved or uneven surfaces
  • Moisture resistant for various light-duty indoor and sheltered outdoor marking needs
  • Square corners and scored back liner for quick peel-off and application
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Specifications
    Material Vinyl
    Mounting Type Self-adhesive
    Shape Rectangular
    Languages English
    Displayed Text NOTICE FOR THE HEALTH AND SAFETY OF ONE ANOTHER FACE MASK REQUIRED
    Color Blue on White