Learn More
Accuform™ Safety Signs
Catalog No. 19169579 Shop All AccuformNMC Products
Change view
Click to view available options
Displayed Text:
CAUTION HAZARDOUS WASTE SATELLITE ACCUMULATION POINT
SATELLITE STORAGE AREA
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text
19-169-579 CAUTION HAZARDOUS WASTE SATELLITE ACCUMULATION POINT
19-169-580 SATELLITE STORAGE AREA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-169-579 Supplier Accuform™ Supplier No. MCHL645VP
Only null left
Add to Cart
Add to Cart
Accuform™ Safety Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage.
  • Durable plastic has slight flex that is ideal for indoor and sheltered outdoor environments
  • Printed surface has clear UV protective overcoat
  • 50% recycled content with UV inhibitors and good impact resistance
  • Rounded corners with 3/16 in. mounting holes for installation
  • Comply in accordance with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
Specifications
Certifications/Compliance OSHA 29, CFR 1910.145, and ANSI Z535
Material Plastic
Mounting Type Corner Holes
Dimensions (L x W) 7 x 10 in.
Displayed Text CAUTION HAZARDOUS WASTE SATELLITE ACCUMULATION POINT
Color Black on Yellow