Learn More
Research Products International Corp Safety-Solve, 1 CS
SDP
Catalog No. 501977049
Encompass_Preferred
Safety-Solve is a ready-to-use high flash point, cocktail for liquid scintillation counting. This low-odor, high flash point, low viscosity formula does not require dilution prior to use. Contains a blend of high quantum efficiency quench resistant fluors in a non-ionic emulsifier system which will accommodate a wide range of samples.
Catalog No. 50-197-7049 Supplier Research Products International Corp Supplier No. 111177CS
Safety-Solve is a ready-to-use high flash point, cocktail for liquid scintillation counting. This low-odor, high flash point, low viscosity formula does not require dilution prior to use. Contains a blend of high quantum efficiency quench resistant fluors in a non-ionic emulsifier system which will accommodate a wide range of samples.