Learn More
Cole-Parmer Safety Spark Laboratory Lighter with 5 Flints
Catalog No. NC1697588 Shop All Cole-Parmer Products
Encompass_Preferred
Use this well-constructed, safety spark lighter for all your burner ignition needs. Special spark hood protects the flint. Gas pocket forms beneath the hood, ensuring instant lighting. Lighter incorporates new larger, renewable flint that produces up to 1500 ignitions..
Catalog No. NC1697588 Supplier Cole-Parmer Supplier No. 3613650
Use this well-constructed, safety spark lighter for all your burner ignition needs. Special spark hood protects the flint. Gas pocket forms beneath the hood, ensuring instant lighting. Lighter incorporates new larger, renewable flint that produces up to 1500 ignitions..