Learn More
GUARDIAN EQUIPMENT Safety Station with Eyewash, Plastic Bowl
Catalog No. NC1146854
Encompass_Preferred
 Combination eyewash and shower safety station. Eyewash features a plastic bowl with two GS-Plus™ spray-type outlet heads that deliver a flood of water for rinsing eyes.
Catalog No. NC1146854 Supplier GUARDIAN EQUIPMENT Supplier No. G1902P
 Combination eyewash and shower safety station. Eyewash features a plastic bowl with two GS-Plus™ spray-type outlet heads that deliver a flood of water for rinsing eyes.