Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Safety Stripe Tape

Catalog No. 19070903 Shop All 3M Products
Change view
Click to view available options
Color:
Black, Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-070-903 Black, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-903 Supplier 3M™ Supplier No. 7000017005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M Company Safety Stripe Tape provides clear safety indication for driving and walking lanes, hazardous areas, protruding equipment and low hanging objects

  • One-piece, clean removal from many surfaces which helps reduce clean-up and labor costs
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Rubber adhesive provides excellent holding strength on many surfaces
  • Good solvent resistance for long service life
  • Quick application of tape eliminates prep work and clean-up processes associated with painting
  • Color is embedded into the tape construction to provide long-term, high visibility and abrasion resistance
  • 3M Catalog# 5702
TRUSTED_SUSTAINABILITY

Specifications

Color Black, Yellow
For Use With (Application) Floor, safety and hazard marking
Packaging Quantity 24/Pk.
Material Vinyl
Type Specialty Tapes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.