Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITL BioMedical Safety SubCulture Unit 2

Catalog No. 22799904 Shop All ITL BioMedical Products
Change view
Click to view available options
Material:
Polycarbonate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
22-799-904 Polycarbonate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-799-904 Supplier ITL BioMedical Supplier No. A100722
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Needleless for a safer subculture technique.

  • Needleless subculture device
  • Simple and safe transfer of sample to slide and media plates
  • Securely fits on a range of blood culture bottles
  • Protective cap captures culture and gas discharge reducing exposure
  • Compliance

    CE marked

    Specifications

    Sterility Sterile
    Color Clear
    Compatibility Designed to fit securely onto a range of culture bottles
    For Use With (Application) Needleless transfer of samples from culture bottles to slides and media plates
    For Use With (Equipment) WASP Copan, Luminex Verigene, Cepheid Xpert, BD Kiestra, Blood Culture Bottles, Media Plates, Microscope slides
    Quantity 48/Pk., 6 Pk./Cs., 288/Cs.
    Material Polycarbonate
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.