Learn More
Sol-Care™ Safety Syringe
Catalog No. 22024427
Change view
Click to view available options
Volume (Metric):
3 mL
5 mL
10 mL
20 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-024-427 20 mL
22-024-411 3 mL
22-024-420 5 mL
22-024-424 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-024-427 Supplier Sol-Care™ Supplier No. 120009IM
Add to Cart
Add to Cart
This item is not returnable. View return policy
With manual retractable needle mechanism, it is compatible with all hypodermic needles. The Sol-Care Safety Syringe aids in the prevention of needlestick injuries by allowing the user to retract the needle into the barrel after medication delivery, covering the needle and locking it in place.
  • Compatible with all standard needles
  • Clear, bold graduations for accurate dosing
  • No Gap design allows the plunger to be pushed to the zero mark without activating the safety mechanism
  • Safety mechanism does not obstruct the view of the needle tip
  • Visual and audible confirmation of safety mechanism activation
Specifications
Autoclavable Not Autoclavable
Material Polypropylene
Needle Gauge No Needle Included
Latex Free Yes
Disposable Single-use
Volume (Metric) 20 mL
Needle Point Style No Needle Included
For Use With (Equipment) Hypodermic Needle, Blunt Needle
Connection Type Luer Lock
Graduated Yes
Sterility Sterile