Learn More
Uline Uline Industrial Vinyl Safety Tape - 3"" x 36 yds, Orange
Catalog No. NC2608199
Encompass_Preferred
Help keep costly accidents down with Uline vinyl safety tape. Industrial 6 mil vinyl with rubber adhesive - unlaminated, general purpose.
Catalog No. NC2608199 Supplier Uline Supplier No. S11644
Help keep costly accidents down with Uline vinyl safety tape. Industrial 6 mil vinyl with rubber adhesive - unlaminated, general purpose.