Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Safety in the Lab Bulletin Board Set of 8
SCI_ED

Catalog No. S43592 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
7 to 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43592 7 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43592 Supplier NewPath Learning™ Supplier No. 945601

missing translation for 'validMainframeMsg'

Perfect for Classroom Reference or Student Activities. NewPath™ Learning Safety in the Lab Bulletin Board Set of 8 covers key concepts on Safety in the Lab.

  • Double-sided 18 x 12 in. chart is laminated
  • One side provides an illustrated overview of the topic, while the reverse features write-on/wipeoff graphic organizers, problem-based questions and other review activities

Includes the following charts:

  1. Lab safety signs and symbols
  2. Personal protective equipment
  3. Safety in the lab
  4. Chemical hazards
  5. Chemical handling
  6. Eye safety
  7. Chemical spills
  8. Biological safety

Specifications

Grade Level Intermediate - Middle School, High School
Grade 7 to 9
Format Chart
Product Type Safety in the Lab Bulletin Board
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.