Learn More
Grainger Conrader Air Safety Valve, Stainless Steel, 150 psi, 1/2 in. (M)NPT, 1 ea.
Catalog No. 502759286
Encompass_Preferred
This hard seat safety valve is engineered for reliable pressure relief in air and gas applications. Designed and certified to ASME Code specifications, the valve features a robust metal-to-metal seal and is suitable for demanding industrial environments.
  • Preset pressure setting of 150 psi
  • 1/2 in. (M)NPT inlet connection
  • Constructed with stainless steel for durability
  • Operating temperature range of -65°F to 400°F
  • Compliant with ASME Boiler and Pressure Vessel Code Section VIII Division 1
  • Certified to meet Pressure Equipment Directive of the European Union with CE mark
  • Flow capacity of 116 SCFM
Catalog No. 50-275-9286 Supplier Grainger Supplier No. 15X855
This hard seat safety valve is engineered for reliable pressure relief in air and gas applications. Designed and certified to ASME Code specifications, the valve features a robust metal-to-metal seal and is suitable for demanding industrial environments.
  • Preset pressure setting of 150 psi
  • 1/2 in. (M)NPT inlet connection
  • Constructed with stainless steel for durability
  • Operating temperature range of -65°F to 400°F
  • Compliant with ASME Boiler and Pressure Vessel Code Section VIII Division 1
  • Certified to meet Pressure Equipment Directive of the European Union with CE mark
  • Flow capacity of 116 SCFM