Learn More
Grainger Conrader Air Safety Valve, Stainless Steel, 1/2 in, 200 psi, 1 ea
Catalog No. 502759732
Encompass_Preferred
The SRH375 is a robust safety valve engineered for air and gas applications. Designed for reliability, this valve is constructed from stainless steel and features a hard seat design to ensure precise performance in high-pressure environments.
  • Preset pressure setting of 200 psi
  • Operating temperature range of -65°F to 400°F
  • 1/2 in. (M)NPT inlet connection
  • Compliant with ASME boiler and pressure vessel code section VIII division 1
  • Features 302 stainless steel spring and 440 stainless steel sealing ball
  • Certified for use under Canada (CRN) standards
  • CE and UK PESR compliant
Catalog No. 50-275-9732 Supplier Grainger Supplier No. 15X857
The SRH375 is a robust safety valve engineered for air and gas applications. Designed for reliability, this valve is constructed from stainless steel and features a hard seat design to ensure precise performance in high-pressure environments.
  • Preset pressure setting of 200 psi
  • Operating temperature range of -65°F to 400°F
  • 1/2 in. (M)NPT inlet connection
  • Compliant with ASME boiler and pressure vessel code section VIII division 1
  • Features 302 stainless steel spring and 440 stainless steel sealing ball
  • Certified for use under Canada (CRN) standards
  • CE and UK PESR compliant