Learn More
Grainger Conrader Safety Valve, Stainless Steel, Hard Seat, 1/2 in. (M)NPT, 275 psi, 1 ea.
Catalog No. 502760436
Encompass_Preferred
The SRH375 is a hard seat safety valve designed for high-temperature air and gas applications, such as intercoolers and after-coolers. This stainless steel valve is precision-engineered to provide reliable pressure relief and is compliant with ASME Boiler and Pressure Vessel Code Section XIII standards.
  • Preset pressure setting of 275 psi.
  • Inlet size of 1/2 in. (M)NPT.
  • Operational temperature range of -65°F to 400°F.
  • Flow rate of 529 SCFM at set pressure.
  • Robust construction featuring 304 stainless steel internal components and a 440 stainless steel ball.
  • Certified to ASME Code specifications with UV and NB symbols.
  • Meets European PED 2014/68/EU and UK PESR requirements.
Catalog No. 50-276-0436 Supplier Grainger Supplier No. 15X859
The SRH375 is a hard seat safety valve designed for high-temperature air and gas applications, such as intercoolers and after-coolers. This stainless steel valve is precision-engineered to provide reliable pressure relief and is compliant with ASME Boiler and Pressure Vessel Code Section XIII standards.
  • Preset pressure setting of 275 psi.
  • Inlet size of 1/2 in. (M)NPT.
  • Operational temperature range of -65°F to 400°F.
  • Flow rate of 529 SCFM at set pressure.
  • Robust construction featuring 304 stainless steel internal components and a 440 stainless steel ball.
  • Certified to ASME Code specifications with UV and NB symbols.
  • Meets European PED 2014/68/EU and UK PESR requirements.