Learn More
Grainger Conrader Hard Seat Pneumatic Safety Valve, Stainless Steel, 1/4 in NPT, 175 psi
Catalog No. 502759283
Encompass_Preferred
This hard seat pneumatic safety valve is designed for overpressure protection in industrial air and gas applications. Featuring a durable metal-to-metal seal, this component provides reliable performance in environments where standard elastomer seals may be unsuitable.
  • Hard seat design for robust performance
  • Stainless steel body, seat, and trim construction
  • Preset relief setting of 175 psi
  • 1/4 in (M)NPT inlet connection
  • Designed and certified to ASME Code specifications
  • Suitable for high-temperature applications up to 400 °F
  • Ideal for corrosive environments
Catalog No. 50-275-9283 Supplier Grainger Supplier No. 15X843
This hard seat pneumatic safety valve is designed for overpressure protection in industrial air and gas applications. Featuring a durable metal-to-metal seal, this component provides reliable performance in environments where standard elastomer seals may be unsuitable.
  • Hard seat design for robust performance
  • Stainless steel body, seat, and trim construction
  • Preset relief setting of 175 psi
  • 1/4 in (M)NPT inlet connection
  • Designed and certified to ASME Code specifications
  • Suitable for high-temperature applications up to 400 °F
  • Ideal for corrosive environments