Learn More
Grainger Conrader Hard Seat Safety Valve, 1/4 in MNPT, 100 psi, 1 ea
Catalog No. 502758866
Encompass_Preferred
This hard seat safety valve is designed for reliable pressure relief in air and gas applications. Featuring a 1/4 in (M)NPT inlet, it comes preset to 100 psi with a broad set pressure range of 25 to 350 psig. The valve is engineered for durability, incorporating high-quality stainless steel components to meet rigorous performance standards.
  • Preset pressure setting of 100 psi
  • Operating temperature range from -65°F to 400°F
  • Flow factor of 0.72 (slope method)
  • Constructed with a 440 stainless steel sealing ball
  • Compliant with ASME Boiler and Pressure Vessel Code Section XIII
  • Suitable for air and gas applications
Catalog No. 50-275-8866 Supplier Grainger Supplier No. 15X840
This hard seat safety valve is designed for reliable pressure relief in air and gas applications. Featuring a 1/4 in (M)NPT inlet, it comes preset to 100 psi with a broad set pressure range of 25 to 350 psig. The valve is engineered for durability, incorporating high-quality stainless steel components to meet rigorous performance standards.
  • Preset pressure setting of 100 psi
  • Operating temperature range from -65°F to 400°F
  • Flow factor of 0.72 (slope method)
  • Constructed with a 440 stainless steel sealing ball
  • Compliant with ASME Boiler and Pressure Vessel Code Section XIII
  • Suitable for air and gas applications