Learn More
Grainger Conrader Safety Valve, Hard Seat, Stainless Steel, 225 psi, 1 ea
Catalog No. 502759284
Encompass_Preferred
This hard seat safety valve is designed for reliable pressure relief in air and gas applications. Featuring a durable stainless steel construction, it is engineered for performance under demanding conditions.
  • Preset pressure setting of 225 psi
  • 1/4 in (M)NPT inlet connection
  • Operating temperature range of -65°F to 400°F
  • Compliant with ASME Boiler and Pressure Vessel Code Section XIII
  • Rated airflow of 188 cfm at set pressure
  • Suitable for air and gas system applications
Catalog No. 50-275-9284 Supplier Grainger Supplier No. 15X845
This hard seat safety valve is designed for reliable pressure relief in air and gas applications. Featuring a durable stainless steel construction, it is engineered for performance under demanding conditions.
  • Preset pressure setting of 225 psi
  • 1/4 in (M)NPT inlet connection
  • Operating temperature range of -65°F to 400°F
  • Compliant with ASME Boiler and Pressure Vessel Code Section XIII
  • Rated airflow of 188 cfm at set pressure
  • Suitable for air and gas system applications