Learn More
Research Products International Corp Safety Vented Wash Bottle, Isopropanol, 16oz, 3/cs
SDP
Catalog No. 501978347
Encompass
Wash Bottles feature patented venting assembly that prevents vapor pressure build-up of solvent within the wash bottle. Eliminates solvent drips from the spout due to internal evaporation. Wash Bottles are labelled with most commonly used solvent names. Bottles are low-density polyethylene, closures and venting assembly are polypropylene.
Catalog No. 50-197-8347 Supplier Research Products International Corp Supplier No. 248674
Wash Bottles feature patented venting assembly that prevents vapor pressure build-up of solvent within the wash bottle. Eliminates solvent drips from the spout due to internal evaporation. Wash Bottles are labelled with most commonly used solvent names. Bottles are low-density polyethylene, closures and venting assembly are polypropylene.