Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Safety-Walk™ Slip-Resistant Conformable Tapes & Treads

Catalog No. 17623859 Shop All 3M Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-859 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-859 Supplier 3M™ Supplier No. 7000051949
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Safety-Walk™ Slip-Resistant Conformable Tapes & Treads are ideal for use on diamond grade safety plates, steps and other irregular surfaces. Mineral coated, slip resistant with aluminum foil backing. Conforms to irregular surfaces and around edges.

  • For irregular or contoured surfaces in light to heavy shoe-traffic areas
  • Mineral-coated, high friction slip-resistant surface
  • Aluminum backing conforms to corners, curves and irregular surfaces like diamond plating, riveted plates and ladder rungs
  • Aggressive, high performance adhesive forms an excellent bond for a secure hold
  • Applications: stairs, ladders, loading ramps, platforms, irregular surfaces like diamond plating or flat surfaces with rivets or screw heads
  • Tapes and treads resist wrinkling, curling, tearing, shrinking, and lifting
  • Safety yellow color for high visibility and easy identification of hazard areas
  • Custom shapes and sizes are also available
  • NFSI High Traction Certification
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NFSI High Traction Certification
Length (Metric) 18.3 m
Temperature Range (Metric) Maximum 79°C
Width (English) 4 in.
Width (Metric) 101.6 mm
Adhesive Type Pressure Sensitive Adhesive
Color Yellow
Type Tape
Length (English) 60 ft.
Packaging Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.