Learn More
Alkali Scientific Safety Wash Bottles, GHS and OSHA Compliant, Self Venting, LDPE, 500mL (16oz), 6/Pk, Distilled Water (Natural)
SDP
Catalog No. 5000662513
Encompass_Preferred
This convenient set includes six 500mL (16oz) Alkali Scientific safety wash bottles, each clearly pre printed with the chemical name and formula, CAS number, and NFPA 704 four color hazard diamond for easy identification and compliance with Right to Know regulations. Product Features: Materials: Bottles are made from translucent low density polyethylene (LDPE); closures and draw tubes are durable polypropylene (PP) Leak Proof Design: Narrow mouth bottles with integrated closure and stem molded as a single piece to prevent leaks and separation during use Flexible Dispensing: Tip can be trimmed to increase flow rate; ideal for short term dispensing Color Coded Caps: Polypropylene screw caps are color coded to assist with quick identification Chemical Compatibility: Includes bottles labeled for Acetone, Ethyl Alcohol, Methanol, Isopropanol, Distilled Water, and Sodium Hypochlorite Applications: Perfect for chemistry labs, biology labs, pharmaceutical settings, and general scientific test
Catalog No. 50-006-62513 Supplier Alkali Scientific Supplier No. SWB00W
This convenient set includes six 500mL (16oz) Alkali Scientific safety wash bottles, each clearly pre printed with the chemical name and formula, CAS number, and NFPA 704 four color hazard diamond for easy identification and compliance with Right to Know regulations. Product Features: Materials: Bottles are made from translucent low density polyethylene (LDPE); closures and draw tubes are durable polypropylene (PP) Leak Proof Design: Narrow mouth bottles with integrated closure and stem molded as a single piece to prevent leaks and separation during use Flexible Dispensing: Tip can be trimmed to increase flow rate; ideal for short term dispensing Color Coded Caps: Polypropylene screw caps are color coded to assist with quick identification Chemical Compatibility: Includes bottles labeled for Acetone, Ethyl Alcohol, Methanol, Isopropanol, Distilled Water, and Sodium Hypochlorite Applications: Perfect for chemistry labs, biology labs, pharmaceutical settings, and general scientific test