Learn More
Sol-Care™ Safety Winged Infusion Set
Catalog No. 22024478
Change view
Click to view available options
Needle Gauge:
21 G
22 G
23 G
25 G
Color:
Black
Dark Blue
Green
Orange
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
22-024-478 25 G Orange
22-024-475 21 G Green
22-024-477 23 G Dark Blue
22-024-476 22 G Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-024-478 Supplier Sol-Care™ Supplier No. 253412SI
Add to Cart
Add to Cart
This item is not returnable. View return policy
Intended for short-term IV medication administration, each infusion set is equipped with a sliding safety mechanism that is activated after the infusion has completed, helping to prevent needlestick injuries.
  • Simple safety mechanism that should be activated immediately after blood withdrawal is complete
  • Reduces the risk of needlestick injury
  • Thumb rest to ease handling during safety activation
  • Flexible wings to secure device to patient's skin
  • Thin walled needle is designed to minimize patient discomfort
  • Available in 21, 23 or 25 gauge
  • Transparent housing and tubing for clear visualization of blood flow
  • DEHP-free, flexible, soft, and kink resistant tubing
Specifications
Color Orange
Material Polyurethane
Needle Gauge 25 G
Sterility Sterile
Latex Free Yes
Length (English) Needle 3/4 in.
Length (English) Tube 12 in.
Length (Metric) Needle 19.05 mm