Learn More
Grainger SafetyInterlockSwitch Plastic 1NO/1NC
Catalog No. 502752690
Encompass_Preferred
The Telemecanique XCSLE-14L755 is a safety interlock switch with a 14mm actuator. It has a plastic housing rated IP67 with 2 NC contacts. This interlock switch is designed for use in machine guarding applications up to PLe/SIL3. It features direct opening operation, tamper resistance, and redundancy in the safety contacts. The operating temperature range is -25°C to +80°C. It requires a minimum door gap of 14mm for proper operation. The switch comes with LED status indicators and has a lifetime up to 1 million operations. The XCSLE series offers various actuator lengths from 14mm up to 110mm.
Catalog No. 50-275-2690 Supplier Grainger Supplier No. 14L754
The Telemecanique XCSLE-14L755 is a safety interlock switch with a 14mm actuator. It has a plastic housing rated IP67 with 2 NC contacts. This interlock switch is designed for use in machine guarding applications up to PLe/SIL3. It features direct opening operation, tamper resistance, and redundancy in the safety contacts. The operating temperature range is -25°C to +80°C. It requires a minimum door gap of 14mm for proper operation. The switch comes with LED status indicators and has a lifetime up to 1 million operations. The XCSLE series offers various actuator lengths from 14mm up to 110mm.