Learn More
Medicom™ SafeWear™ Hipster Jacket™, Poppy Pink
Catalog No. 19910770 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-770 X-Large
19-910-767 Small
19-910-768 Medium
19-910-769 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-910-770 Supplier Medicom™ Supplier No. 8116D
  • $301.00
    $244.65 / Case of 60
    Save $56.35
    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
The SafeWear™ Hipster Jacket™, Poppy Pink is premium protective apparel made from top-quality materials and designed to provide reliable barrier protection without sacrificing comfort or style.
  • Available in a wide range of styles, colours and sizes
  • Comply with recommended OSHA best practices
  • Made from high-quality 45 g/m2 SMS fabric
  • Detailed finishing, 3-pocket design and front snap closure
  • Each bag contains 10+2 units, for a total of 12 vs. 10 per package from the competition
  • Fluid-resistant fabric meets AAMI PB70 Level 2 standards
  • Fluid-resistant fabric meets AAMI PB70 Level 2 standards
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material SMS Polypropylene
Color Poppy Pink
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Fluids
No. of Pockets 3
Latex Free Yes