Learn More
Medicom™ SafeWear™ Hipster Jacket™, White
Catalog No. 19910762 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-762 X-Large
19-910-761 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-910-762 Supplier Medicom™ Supplier No. 8105D
Add to Cart
Add to Cart
This item is not returnable. View return policy
The SafeWear™ Hipster Jacket™, White is premium protective apparel made from top-quality materials and designed to provide reliable barrier protection without sacrificing comfort or style.
  • Available in a wide range of styles, colours and sizes
  • Comply with recommended OSHA best practices
  • Made from high-quality 45 g/m2 SMS fabric
  • Detailed finishing, 3-pocket design and front snap closure
  • Each bag contains 10+2 units, for a total of 12 vs. 10 per package from the competition
  • Fluid-resistant fabric meets AAMI PB70 Level 2 standards
  • Fluid-resistant fabric meets AAMI PB70 Level 2 standards
Specifications
Product Type Single-use Lab Coat
Size X-Large
Gender Unisex
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Latex Free Yes