Learn More
Medicom™ SafeWear™ SMS Hipster Jacket Lab Coat
Catalog No. 19910831 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Color:
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-910-831 Small White
19-910-832 Medium White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-910-831 Supplier Medicom™ Supplier No. 8105A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Procedure SMS Jacket designed to protect health professionals and patients from cross contamination. Single use and disposable gowns made of polypropylene. Material meets AAMI/PB70 Level 2 requirements for impact penetration and hydrostatic pressure. Latex free
  • High quality ultra-soft & light weight SMS fabric (Weight 45 g/m2 for Jacket & Lab Coat)
  • Fluid-resistant material meets AAMI Level 2
  • Designed to fit properly with knitted cuffs & collar
  • High-quality front snap closure buttons
  • Available in a wide range of sizes and in fashionable colors
  • 12 units in every pack
Specifications
Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Style Hipster Jacket
No. of Pockets 3
Latex Free Yes
Autoclavable Not Autoclavable