Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safflower oil, Food grade, MP Biomedicals™

Catalog No. ICN10288890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10288890 500 mL
ICN10288891 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10288890 Supplier MP Biomedicals, Inc Supplier No. 0210288890
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8001-23-8

Specifications

Color Yellow
Quantity 500 mL
Grade Food
Physical Form Oil
Chemical Name or Material Safflower oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.