Learn More
Saffron, Sigma-Aldrich
Catalog No. 01258824 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
01-258-824 5 g
1 options
Catalog No. 01-258-824 Supplier MilliporeSigma Supelco Supplier No. S83815G
Add to Cart
Add to Cart
Dried flower stigmas suitable for use in histological staining procedures.
Saffron is a crude source of the carotenoid antioxidants crocein and crocetin, as well as other carotenoids and monoterpene aldehydes. Saffron has been studied for use as an anticancer and chemopreventive agent and for its effects on antioxidant defense system pathways. A study in mice reported treatment with saffron reduced lipid peroxidation and caspase-3 activity, while increasing total brain antioxidant activity.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 g
Product Type Stain Kit