Learn More
Cayman Chemical Safinamide Metabolite NW 1689, Purity: ≥95%, CAS: 405-85-6, 100 mg
SDP
Catalog No. 502913785
Encompass_Preferred
A potential impurity found in commercial preparations of safinamide; also a degradation product of safinamide formed under conditions of thermal and oxidative stress
Catalog No. 50-291-3785 Supplier Cayman Chemical Supplier No. 35395100 MG
A potential impurity found in commercial preparations of safinamide; also a degradation product of safinamide formed under conditions of thermal and oxidative stress