Learn More
Cayman Chemical Safinamide Acid, Purity: ≥95%, CAS: 1160513-60-9, 10 mg
SDP
Catalog No. 502910340
Encompass_Preferred
A potential impurity found in commercial preparations of safinamide; a degradation product formed under conditions of thermal and oxidative stress
Catalog No. 50-291-0340 Supplier Cayman Chemical Supplier No. 3508410 MG
A potential impurity found in commercial preparations of safinamide; a degradation product formed under conditions of thermal and oxidative stress