Learn More
Medchemexpress LLC Safinamide acid | 1160513-60-9 | 99.5% | 303.33 g/mol | C17H18FNO3 | 25mg
SDP
Catalog No. 5000269136
Encompass_Preferred
Safinamide acid (Compound Imp-E) is the metabolite of Safinamide (HY-70057) Safinamide acid can be formed by hydrolysis of Safinamide intermediate under alkaline or acidic conditions Safinamide acid is utilized to monitor process-related impurities and degradation products in safinamide samples[1]
Catalog No. 50-002-69136 Supplier Medchemexpress LLC Supplier No. HYW71754825MG
Add to Cart
Add to Cart
Safinamide acid (Compound Imp-E) is the metabolite of Safinamide (HY-70057) Safinamide acid can be formed by hydrolysis of Safinamide intermediate under alkaline or acidic conditions Safinamide acid is utilized to monitor process-related impurities and degradation products in safinamide samples[1]