Learn More
Medchemexpress LLC Safinamide-d4-1 | 2748522-33-8 | 98.9% | C17H15D4FN2O2 | 1 MG
SDP
Catalog No. 5000434350
Encompass_Preferred
Safinamide-d4-1 is deuterium labeled Safinamide, a potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor. It also blocks sodium channels and modulates glutamate release. Safinamide-d4-1 has neuroprotective and neurorescuing effects, making it suitable for research into Parkinson's disease and ischemic stroke. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor
  • Blocks sodium channels and modulates glutamate release
  • Offers neuroprotective and neurorescuing effects
Catalog No. 50-004-34350 Supplier Medchemexpress LLC Supplier No. HY70057S11MG
Safinamide-d4-1 is deuterium labeled Safinamide, a potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor. It also blocks sodium channels and modulates glutamate release. Safinamide-d4-1 has neuroprotective and neurorescuing effects, making it suitable for research into Parkinson's disease and ischemic stroke. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor
  • Blocks sodium channels and modulates glutamate release
  • Offers neuroprotective and neurorescuing effects