Learn More
Medchemexpress LLC Safinamide mesylate | 202825-46-5 | 99.07% | C18H23FN2O5S | 1 ML
SDP
Catalog No. 5000433687
Encompass_Preferred
Safinamide mesylate is a potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor, blocking sodium channels and modulating glutamate (Glu) release. It demonstrates neuroprotective and neurorescuing effects, making it suitable for the study of Parkinson's disease and ischemic stroke.
  • Potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor
  • Blocks sodium channels
  • Modulates glutamate (Glu) release
  • Demonstrates neuroprotective and neurorescuing effects
  • Used for the study of Parkinson's disease
  • Used for the study of ischemic stroke
Catalog No. 50-004-33687 Supplier Medchemexpress LLC Supplier No. HY70057A10MM/1ML
Safinamide mesylate is a potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor, blocking sodium channels and modulating glutamate (Glu) release. It demonstrates neuroprotective and neurorescuing effects, making it suitable for the study of Parkinson's disease and ischemic stroke.
  • Potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor
  • Blocks sodium channels
  • Modulates glutamate (Glu) release
  • Demonstrates neuroprotective and neurorescuing effects
  • Used for the study of Parkinson's disease
  • Used for the study of ischemic stroke