Learn More
Medchemexpress LLC Safinamide mesylate | 202825-46-5 | 99.1% | C18H23FN2O5S | 5 MG
SDP
Catalog No. 5000433641
Encompass_Preferred
Safinamide mesylate is a potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor (IC50=0.098 μM) over MAO-A (IC50=580 nM). It also blocks sodium channels and modulates glutamate (Glu) release, showing greater affinity at depolarized (IC50=8 μM) than at resting (IC50=262 μM) potentials. It exhibits neuroprotective and neurorescuing effects and can be used for the study of Parkinson's disease, ischemic stroke, and other conditions.
  • Potent, selective, and reversible monoamine oxidase B inhibitor
  • Blocks sodium channels
  • Modulates glutamate release
  • Exhibits neuroprotective and neurorescuing effects
  • Suitable for the study of Parkinson's disease and ischemic stroke
Catalog No. 50-004-33641 Supplier Medchemexpress LLC Supplier No. HY70057A5MG
Safinamide mesylate is a potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor (IC50=0.098 μM) over MAO-A (IC50=580 nM). It also blocks sodium channels and modulates glutamate (Glu) release, showing greater affinity at depolarized (IC50=8 μM) than at resting (IC50=262 μM) potentials. It exhibits neuroprotective and neurorescuing effects and can be used for the study of Parkinson's disease, ischemic stroke, and other conditions.
  • Potent, selective, and reversible monoamine oxidase B inhibitor
  • Blocks sodium channels
  • Modulates glutamate release
  • Exhibits neuroprotective and neurorescuing effects
  • Suitable for the study of Parkinson's disease and ischemic stroke