Learn More
TARGETMOL CHEMICALS INC SAFINAMIDE MESYLATE 25MG
SDP
Catalog No. 502579318
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Safinamide mesylate (EMD 1195686 mesylate) a mesylate salt of Safinamide can reversibly and specifically inhibit MAO-B (IC50 98 nM) has 5918-fold selectivity against MAO-A. purity: 98%
Catalog No. 50-257-9318 Supplier TARGETMOL CHEMICALS INC Supplier No. T665125MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Safinamide mesylate (EMD 1195686 mesylate) a mesylate salt of Safinamide can reversibly and specifically inhibit MAO-B (IC50 98 nM) has 5918-fold selectivity against MAO-A. purity: 98%