Learn More
Selleck Chemical LLC Safinamide Mesylate S1472-5mg
SDP
Catalog No. 503110622
Encompass_Preferred
Safinamide Mesylate (PNU-151774E FCE28073) is mesylate salt of Safinamide selectively and reversibly inhibits MAO-B with IC50 of 98 nM exhibits 5918-fold selectivity against MAO-A Phase 3
Catalog No. 50-311-0622 Supplier Selleck Chemical LLC Supplier No. S14725MG
Add to Cart
Add to Cart
Safinamide Mesylate (PNU-151774E FCE28073) is mesylate salt of Safinamide selectively and reversibly inhibits MAO-B with IC50 of 98 nM exhibits 5918-fold selectivity against MAO-A Phase 3