Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safinamide 98.0+%, TCI America™
SDP

Catalog No. S0935100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0935100MG 100 mg
S093525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0935100MG Supplier TCI America Supplier No. S0935100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133865-89-1
Molecular Formula C17H19FN2O2
Molecular Weight (g/mol) 302.349
MDL Number MFCD00897036
InChI Key NEMGRZFTLSKBAP-LBPRGKRZSA-N
Synonym (S)-2-[[4-[(3-Fluorobenzyl)oxy]benzyl]amino]propionamide
PubChem CID 131682
IUPAC Name (2S)-2-[[4-[(3-fluorophenyl)methoxy]phenyl]methylamino]propanamide
SMILES CC(C(=O)N)NCC1=CC=C(C=C1)OCC2=CC(=CC=C2)F

Specifications

Melting Point 212°C
Color White
Quantity 100 mg
Formula Weight 302.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Safinamide
RTECSNumber TX1457385
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.