Learn More
Cayman Chemical Safinamide (mesylate), Purity: ≥98%, CAS: 202825-46-5, 10 mg
SDP
Catalog No. 502986503
Encompass_Preferred
An inhibitor of MAO-B (IC50 = ~0.1 μM); selective for MAO-B over MAO-A (IC50 = >10 μM); inhibits radioligand binding to sodium channel binding site 2, sigma-1, and sigma-2 receptors in rat brain membranes (IC50s = 8.2, 0.019, and 1.59 μM, respectively); inhibits high voltage-activated calcium currents and depolarization-induced tetrodotoxin-sensitive fast sodium currents in rat hippocampal neurons; inhibits veratrine-induced glutamate release in rat hippocampal slices (IC50 = 56 μM); inhibits maximal electroshock-induced tonic extension seizures in mice and rats (ED50s = 8 and 11.8 mg/kg, p.o.) as well as maximal seizures induced by bicuculline, picrotoxin, 3-mercaptopropionic acid, and strychnine in mice (ED50s = 26.9, 60.6, 21.5, and 104.1 mg/kg, p.o., respectively)
Catalog No. 50-298-6503 Supplier Cayman Chemical Supplier No. 2154610 MG
An inhibitor of MAO-B (IC50 = ~0.1 μM); selective for MAO-B over MAO-A (IC50 = >10 μM); inhibits radioligand binding to sodium channel binding site 2, sigma-1, and sigma-2 receptors in rat brain membranes (IC50s = 8.2, 0.019, and 1.59 μM, respectively); inhibits high voltage-activated calcium currents and depolarization-induced tetrodotoxin-sensitive fast sodium currents in rat hippocampal neurons; inhibits veratrine-induced glutamate release in rat hippocampal slices (IC50 = 56 μM); inhibits maximal electroshock-induced tonic extension seizures in mice and rats (ED50s = 8 and 11.8 mg/kg, p.o.) as well as maximal seizures induced by bicuculline, picrotoxin, 3-mercaptopropionic acid, and strychnine in mice (ED50s = 26.9, 60.6, 21.5, and 104.1 mg/kg, p.o., respectively)