Learn More
TARGETMOL CHEMICALS INC Safingol hydrochloride 1MG
SDP
Catalog No. 502876766
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Safingol hydrochloride (L-threo-dihydrosphingosine hydrochloride) is a specific inhibitor of protein kinase C. It induces autophagy in solid tumor cells and cancer cell death by inhibiting the PKC and PI3-kinase pathways.BIIB 722 Mesylate is a selective sodium-hydrogen exchange inhibitor. Safingol hydrochloride inhibits PKC and PI3k. Purity 99.71%
Catalog No. 50-287-6766 Supplier TARGETMOL CHEMICALS INC Supplier No. T839781MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Safingol hydrochloride (L-threo-dihydrosphingosine hydrochloride) is a specific inhibitor of protein kinase C. It induces autophagy in solid tumor cells and cancer cell death by inhibiting the PKC and PI3-kinase pathways.BIIB 722 Mesylate is a selective sodium-hydrogen exchange inhibitor. Safingol hydrochloride inhibits PKC and PI3k. Purity 99.71%