Learn More
Electron Microscopy Sciences Safranin 0, 0.1% 500 ML
SDP
Catalog No. 5030115
Encompass_Preferred
Used in Weigert's Iron Hematoxylinwith Methachromic Dyes for nuclear stain. See Weigert, K., Z. Wissen. Mikro., 21:1-5, 1904, Lillie, R.D., Histopathologic Technical and Practical Histochemistry, 3rd edition, McGraw-Hill, NY, 1965, or Clark, G., (ed.), Staining Procedures, 3rd Edition, Williams & Wilkins, Baltimore, p.40, c 1973.
Catalog No. 50-301-15 Supplier Electron Microscopy Sciences Supplier No. 2675805
Used in Weigert's Iron Hematoxylinwith Methachromic Dyes for nuclear stain. See Weigert, K., Z. Wissen. Mikro., 21:1-5, 1904, Lillie, R.D., Histopathologic Technical and Practical Histochemistry, 3rd edition, McGraw-Hill, NY, 1965, or Clark, G., (ed.), Staining Procedures, 3rd Edition, Williams & Wilkins, Baltimore, p.40, c 1973.