Learn More
Electron Microscopy Sciences Safranin O 0.2% 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2617003
Encompass_Preferred
Used in Prussian Blue Method for Hemosiderin, Lillie (1965); Highman (1962); Perls (1867). References are Perls, M., Virchow's Arch. F. Path. Anat., 39:42-48, 1867, Highman, B., Arch. Path., 33:937-938, 1942, Lillie, R.D., Histopathologic Technic & Practical Histochemistry, 3rd Ed., McGraw Hill, N.Y., 1965, and Clark, G., (ed.), Staining Procedures, 3rd Ed., Williams & Wilkins, Baltimore, p. 157, c. 1973.
Catalog No. 50-317-14